Anti-ACTN1 Mouse Monoclonal Antibody [clone: 3F1]

Supplier: Abnova
H00000087-M01A
10615-562EA 335 USD
10615-562
Anti-ACTN1 Mouse Monoclonal Antibody [clone: 3F1]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant ACTN1.

Type: Primary
Antigen: ACTN1
Clonality: Monoclonal
Clone: 3F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR