Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Surgi-OR™ Rochester-Ochsner Forceps, Mid-Grade, Sklar

Supplier: Sklar
Surgi-OR™
95-500 95-512 95-501 95-513 95-514 95-525 95-521 95-504 95-515 95-516 95-517 95-518
Surgi-OR™ Rochester-Oschner Forceps are a heavy instrument designed to grasp medium to heavy tissue or occlude heavy, dense vessels.

They have horizontal serrations the entire length of the jaw as well as 1 x 2 teeth at the tip. The combination of full serrations plus teeth ensure a firm grip on the tissue or vessel being held. Rochester-Oschner Forceps are available either curved or straight and in multiple lengths.

Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.



 

ORDER

Expand All / Collapse All
Material Length Jaw Type Blade Type Supplier No. VWR Catalog Number Unit Price Quantity
Stainless Steel 140 mm (5 1/2") Serrated Straight 95-500 89023-652 Each Retrieving Restricted
Stainless Steel 140 mm (5 1/2") Serrated Curved 95-501 89023-654 Each Retrieving Restricted
Stainless Steel 159 mm (6 1/4") Serrated Straight 95-504 89023-656 Each Retrieving Restricted
Stainless Steel 159 mm (6 1/4") Serrated Curved 95-512 89023-658 Each Retrieving Restricted
Stainless Steel 184 mm (7 1/4") Serrated Straight 95-521 89023-672 Each Retrieving Restricted
Stainless Steel 184 mm (7 1/4") Serrated Curved 95-525 89023-674 Each Retrieving Restricted
Stainless Steel 203 mm (8") Serrated Straight 95-513 89023-660 Each Retrieving Restricted
Stainless Steel 203 mm (8") Serrated Curved 95-514 89023-662 Each Retrieving Restricted
Stainless Steel 229 mm (9") Serrated Straight 95-515 89023-664 Each Retrieving Restricted
Stainless Steel 229 mm (9") Serrated Curved 95-516 89023-666 Each Retrieving Restricted
Stainless Steel 254 mm (10") Serrated Straight 95-517 89023-668 Each Retrieving Restricted
Stainless Steel 254 mm (10") Serrated Curved 95-518 89023-670 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty