Would you like to log in again?
Undercounter Auto Defrost Freezer with Solid Door, Nor-Lake® Scientific
Supplier: NOR-LAKE SCIENTIFIC
NSPF041WMW/0
The Nor-Lake® Scientific Undercounter Auto Defrost Freezer features a digital LED display with microprocessor temperature controller.
  • The display provides continuous product temperature
  • Audible and visual Hi/Lo temperature alarm
  • Unit offers adjustable temperature control from –15 to –35°C, but are supplied factory preset at –20°C
The exterior cabinet front, sides, and top are painted white
Additional standard features include a solid base with leveler legs, one coated wire shelf, air cooled condensing unit, base mounted forced air front-breathing refrigeration system, automatic defrost and condensate evaporation, remote alarm contacts, product sensor with glycerin bottle, sensor access port (1" dia.), condenser filter screen for easy maintenance, and a field reversible door. Air intake and exhaust through the base of the unit for build-in on three sides.
The exterior cabinet back and bottom are galvanized steel. The interior cabinet liner and interior door are smooth aluminum. Cabinet and solid door are foamed-in-place with EPA-compliant, high-density polyurethane foam insulation. The heavy-duty solid door features self-closing hinges, a pull door handle, magnetic door gasket, and centered key door lock (with 2 keys).
Certifications: UL, C-UL listed.
Delivery information: Units are supplied with an 18-month parts and labor warranty, and a 5-year compressor warranty (US and Canada), or a 18-month parts warranty (International).
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Electrical Volume Dimensions No. of Doors Supplier No. VWR Catalog Number Unit Price Quantity
Undercounter Auto Defrost Freezer with Solid Door 115V, 1PH 119 L (4.2 cu. ft.) 60.6W x 80.3H x 63.5D cm (23 7/8 x 31 5/8 x 25") 1 Door NSPF041WMW/0 10799-024 Each Retrieving
UL Listed cUL Listed
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty