Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Precision pH Buffers, Ricca Chemical Company

Supplier: Ricca Chemical
R1552000-500G R1602000-500G R1502000-500G
This precision line of pH Buffers offers tolerances of ±0.005 pH unit or better

Specially selected containers and proprietary manufacturing processes help create some of the most accurate buffers on the market.

Certifications: Compared against and certified traceable to NIST Standard Reference Materials.




 

ORDER

Expand All / Collapse All
Size pH Value Color pH Accuracy Packed Reference Temperature Supplier No. VWR Catalog Number Unit Price Quantity
500 mL 4.00 Red ±0.002 Poly Bottle 25 °C R1502000-500G RC1502-16 Case of 12 Retrieving Restricted
Each Retrieving Restricted
500 mL 7.000 Yellow ±0.002 Poly Bottle 25 °C R1552000-500G RC1552-16 Case of 12 (500 ml) Retrieving Restricted
Each(500ml) Retrieving Restricted
500 mL 10.00 Blue ±0.005 Poly Bottle 25 °C R1602000-500G RC160216 Case of 12 (500ml) Retrieving Restricted
Each (500ml) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty