Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Ala-Tyr-Pro-Gly-Lys-Phe-NH₂, Bachem

Supplier: BACHEM AMERICAS INC
H-6046.0001
The peptide AYPGKF-amide based on the proteolytically-revealed tethered ligand.

  • SequenceSequence: H-Ala-Tyr-Pro-Gly-Lys-Phe-NH₂
  • CAS-No.: 352017-71-1
  • Mol.weight: 680.80
  • SumFormula: C₃₄H₄₈N₈O₇
  • Bulk item no.: H-6046
  • Synonym(s): AYPGKFamide#(Ala¹)-Proteinase Activated Receptor 4 (1-6) amide (mouse)#(Ala¹)-Thrombin Receptor-Like 3 (1-6) amide (mouse)#(Ala¹)-Coagulation Factor II Receptor-Like 3 (1-6) amide (mouse)#AYPGKF-NH2 Selective Protease-Activated Receptor 4 (PAR4) Agonist
  • EINECS no.: -
  • MDL no.: MFCD04974484

PAR peptides (TRAP peptides and thrombin receptor-like peptides) activate the proteinase-activated receptors PAR-1 to PAR-4.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 mg H-6046.0001 H-6046.0001BA Each (1mg) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Aypgkfamide
CAS No. 352017-71-1
Molecular Weight 680.8
Molecular Formula C₃₄H₄₈N₈O₇
Storage Conditions -20 ± 5 °C
Salt Trifluoroacetate
Peptide Aypgkfamide
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty