Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Diaminopimelic acid, Bachem

Supplier: BACHEM AMERICAS INC
F-1505.0001
A non-competitive inhibitor of aspartokinase I, corresponds to ε-carboxylysine.

  • Sequence: 2,6-Diaminopimelic acid
  • CAS-No.: 583-93-7
  • Mol.weight: 190.20
  • SumFormula: C₇H₁₄N₂O₄
  • Bulk item no.: F-1505
  • Synonym(s): 2,6-Diaminoheptanedioic acid
  • EINECS no.: 209-524-6
  • MDL no.: MFCD00002637




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 g F-1505.0001 F-1505.0001BA Each (1g) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name 2 6-Diaminopimelic Acid
CAS No. 583-93-7
Molecular Weight 190.2
Molecular Formula C₇H₁₄N₂O₄
Storage Conditions Room Temperature
Binding (Mixture of LL-, DD- and meso-isomers)
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty