Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Tyr-OMe, Bachem

Supplier: BACHEM AMERICAS INC
E-2750.0005
  • Sequence: H-Tyr-OMe
  • CAS-No.: 1080-06-4
  • Mol.weight: 195.22
  • SumFormula: C₁₀H₁₃NO₃
  • Bulk item no.: E-2750
  • Synonym(s):
  • EINECS no.: 214-095-3
  • MDL no.: MFCD00002392

Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine.Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
5 g E-2750.0005 E-2750.0005BA Each (5g) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name H-Tyr-Ome
CAS No. 1080-06-4
Molecular Weight 195.22
Molecular Formula C₁₀H₁₃NO₃
Storage Conditions -20 ± 5 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty