Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Ala-Leu-Cys-Asn-Cys-Asn-Arg-Ile-Ile-Ile-Pro-His-Gln-Cys-Trp-Lys-Lys-Cys-Gly-Lys-Lys-NH₂, Bachem

Supplier: BACHEM AMERICAS INC
N-2005.1000 N-2005.0500
Tertiapin Q (TPN Q) shows the same channel-blocking activity as the bee venom peptide tertiapin (N-1745), but increased stability due to the exchange of Met¹³ by Gln.

  • Sequence: H-Ala-Leu-Cys-Asn-Cys-Asn-Arg-Ile-Ile-Ile-Pro-His-Gln-Cys-Trp-Lys-Lys-Cys-Gly-Lys-Lys-NH₂
  • CAS-No.: 252198-49-5
  • Mol.weight: 2452.04
  • SumFormula: C₁₀₆H₁₇₅N₃₅O₂₄S₄
  • Bulk item no.: N-2005
  • Synonym(s): TPNQ#Tertiapin (M13Q)#(Gln¹³)-Tertiapin
  • EINECS no.: -
  • MDL no.: MFCD11045948




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.5 mg N-2005.0500 N-2005.0500BA Each (500µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

1 mg N-2005.1000 N-2005.1000BA Each (1mg) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Tertiapin-Q
CAS No. 252198-49-5
Molecular Weight 2452.04
Molecular Formula C₁₀₆H₁₇₅N₃₅O₂₄S₄
Storage Conditions -20 ± 5 °C
Salt Trifluoroacetate
Binding (Disulfide bonds between Cys³ and Cys¹⁴/Cys⁵ and Cys¹⁸)
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty