beta-Amyloid (1-39) Trifluoroacetate
Supplier: Bachem
4073896.1000 4073896.0500
10796-788EA 316.69 USD
10796-788 10796-790
beta-Amyloid (1-39) Trifluoroacetate
Proteins and Peptides
Small quantities of Aβ37, 38 and 39 can be detected in CSF together with Aβ40, the most abundant Aβ homolog, Aβ42, and N-terminally truncated amyloid peptides. The relative amounts depend on the variant of Alzheimer's disease. The C-terminally truncated amyloid peptides are also found in amyloid plaques.