Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Ser-Lys-Lys-Lys-Lys-Lys-Lys-NH₂, Bachem

Supplier: BACHEM AMERICAS INC
H-7426.0500 H-7426.1000
Lixisenatide is an exenatide-related GLP-1 agonist injected once daily for glycemic control in adult diabetes II. In myocardial ischemia-reperfusion injury studies with rats, a cardioprotective activity could be shown. Furthermore, administration of lixisenatide rescued spatial memory and synaptic plasticity from amyloid β-protein-induced impairments. Offered under Bolar Exemption: All APIs that are sold for development of drug products still patent protected are offered under Bolar Exemption only. The following disclaimer applies: These products are offered and sold in small quantities only and solely for uses reasonably related to privileged trials and studies for obtaining marketing authorization required by law (Bolar Exemption). Bachem cannot be made liable for any infringement of intellectual property rights. It is the sole and only responsibility of the purchaser or user of these products to comply with the relevant national rules and regulations.

  • Sequence: H-His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Ser-Lys-Lys-Lys-Lys-Lys-Lys-NH₂
  • CAS-No.: 320367-13-3
  • Mol.weight: 4858.56
  • SumFormula: C₂₁₅H₃₄₇N₆₁O₆₅S
  • Bulk item no.: H-7426
  • Synonym(s): (Des-Pro³⁸)-Exendin-4(-Lys)₆ amide#ZP10
  • EINECS no.: -
  • MDL no.:




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.5 mg H-7426.0500 10796-756 Each (500µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

1 mg H-7426.1000 10796-758 Each (1mg) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Lixisenatide H-7426.1000 Lixisenatide H-7426.0500
CAS No. 320367-13-3
Molecular Weight 4858.56
Molecular Formula C₂₁₅H₃₄₇N₆₁O₆₅S
Storage Conditions -20 ± 5 °C
Salt Acetate
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty