Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Ala-Arg-Val-Tyr-Ile-His-Pro-OH, Bachem

Supplier: BACHEM AMERICAS INC
H-7424.0025 H-7424.0010
Alamandine can be formed from angiotensin A by action of ACE-2 or directly from angiotensin-(1-7) by decarboxylation of its aspartate residue. The angiotensin A analog produces effects resembling those of Ang II (1-7). However, it acts independently of the two known vasodilators receptors of the RAS (Mas and angiotensin II type 2). To produce its effects, alamandine binds to the Mas-related receptor, MrgD. A novel orally active formulation of alamandine produced a long-term antihypertensive effect in spontaneously hypertensive rats and cardioprotective effects. These novel findings will be helpful for developing a new understanding of the RAS, a key regulator of blood pressure and fluid balance. The heptapeptide could serve as a model peptide, e.g. in the development and evaluation of analytical methods.

  • Sequence: H-Ala-Arg-Val-Tyr-Ile-His-Pro-OH
  • CAS-No.: 1176306-10-7
  • Mol.weight: 855.01
  • SumFormula: C₄₀H₆₂N₁₂O₉
  • Bulk item no.: H-7424
  • Synonym(s): Alamandine#(Ala¹)-Angiotensin I/II (1-7)
  • EINECS no.: -
  • MDL no.:

The sequences given for angiotensin I (H-1680) and II (H-1705) correspond to the sequences of human, horse, sheep, pig, mouse, and rat angiotensins I and II.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
10 mg H-7424.0010 10796-724 Each (10mg) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

25 mg H-7424.0025 10796-726 Each (25mg) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Angiotensin A (1-7)
CAS No. 1176306-10-7
Molecular Weight 855.01
Molecular Formula C₄₀H₆₂N₁₂O₉
Storage Conditions -20 ± 5 °C
Salt Trifluoroacetate

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty