Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Val-Met-Val-Gly-Gly-Val-Val-OH, Bachem

Supplier: BACHEM AMERICAS INC
Cerebral amyloid angiopathy (CAA) is a common finding in Alzheimer’s disease in which amyloid-Aβ vascular deposits are featured in 80% of the cases. Mutations in the positions 21-23 (e.g. Dutch mutation E22Q) are primarily associated with CAA, although they manifest with strikingly different clinical phenotypes: cerebral hemorrhage or dementia. The Piedmont L34V Aβ mutant, located outside this hot spot, shows a similar hemorrhagic phenotype, albeit less aggressive than the widely studied Dutch variant.

  • Sequence: H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Val-Met-Val-Gly-Gly-Val-Val-OH
  • CAS-No.: 1678415-83-2
  • Mol.weight: 4315.84
  • SumFormula: C₁₉₃H₂₉₃N₅₃O₅₈S
  • Bulk item no.: H-7414
  • Synonym(s): Amyloid β-Protein (1-40) L34V#Amyloid β-Protein (1-40) Piedmont Mutation
  • EINECS no.:
  • MDL no.:



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.5 mg H-7414.0500 10796-822 Each (500µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

1 mg H-7414.1000 10796-824 Each (1mg) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name (Val34)-beta-Amyloid
CAS No. 1678415-83-2
Molecular Weight 4315.84
Molecular Formula C₁₉₃H₂₉₃N₅₃O₅₈S
Storage Conditions -20 ± 5 °C
Salt Trifluoroacetate
Peptide (Val34)-beta-Amyloid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty