Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fluorescein-5-carbonyl-Lys-Cys-Asn-Thr-Ala-Thr-Cys-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-Arg-Ser-Ser-Asn-Asn-Leu-Gly-Pro-Val-Leu-Pro-Pro-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-NH₂, Bachem

Supplier: BACHEM AMERICAS INC
H-7396.0500 H-7396.1000
  • Sequence: Fluorescein-5-carbonyl-Lys-Cys-Asn-Thr-Ala-Thr-Cys-Ala-Thr-Gln-Arg-Leu-Ala-Asn-Phe-Leu-Val-Arg-Ser-Ser-Asn-Asn-Leu-Gly-Pro-Val-Leu-Pro-Pro-Thr-Asn-Val-Gly-Ser-Asn-Thr-Tyr-NH₂
  • CAS-No.: 1678415-03-6
  • Mol.weight: 4278.75
  • SumFormula: C₁₈₈H₂₈₂N₅₂O₅₉S₂
  • Bulk item no.: H-7396
  • Synonym(s):
  • EINECS no.: -
  • MDL no.:




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.5 mg H-7396.0500 10796-818 Each (500µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

1 mg H-7396.1000 10796-820 Each (1mg) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name 5-Fam-Amylin
Species Mouse
CAS No. 1678415-03-6
Molecular Weight 4278.75
Molecular Formula C₁₈₈H₂₈₂N₅₂O₅₉S₂
Storage Conditions -20 ± 5 °C
Salt Trifluoroacetate
Binding (Disulfide bond)
Peptide 5-Fam-Amylin
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty