Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fmoc-Ala-Ser(Psi(Me,Me)pro)-OH, Bachem

Supplier: BACHEM AMERICAS INC
  • Sequence: Fmoc-Ala-Ser(Psi(Me,Me)pro)-OH
  • CAS-No.: 252554-78-2
  • Mol.weight: 438.48
  • SumFormula: C₂₄H₂₆N₂O₆
  • Bulk item no.: B-4270
  • Synonym(s):
  • EINECS no.: -
  • MDL no.:

These dipeptide building blocks containing Ser- or Thr-derived oxazolidines (pseudoprolines) proved to be versatile tools for overcoming some intrinsic problems in the field of peptide chemistry. The presence of pseudoprolines within a peptide sequence results in the disruption of β-sheet structures considered as a source of intermolecular aggregation during chain elongation, thus increasing solvation and coupling kinetics in peptide assembly. Therefore, use of pseudoprolines offer new possibilities for accessing large peptides by convergent strategies and chemoselective ligation techniques. Moreover, incorporation of a pseudoproline unit facilitates cyclization of peptides.



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 g B-4270.0001 B-4270.0001BA Each (1g) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

5 g B-4270.0005 B-4270.0005BA Each (5g) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Fmoc-Ala-Ser(Psi(Me,Me)Pro)-Oh
CAS No. 252554-78-2
Molecular Weight 438.48
Molecular Formula C₂₄H₂₆N₂O₆
Peptide Fmoc-Ala-Ser(Psi(Me,Me)Pro)-Oh
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty