Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ac-D-2-Nal-D-4-Cpa-D-3-Pal-Ser-N-Me-Tyr-D-Hci-Nle-Arg-Pro-D-Ala-NH₂, Bachem

Supplier: BACHEM AMERICAS INC
H-7384.0001 H-7384.0005
GnRH antagonist. Offered under Bolar Exemption: All APIs that are sold for development of drug products still patent protected are offered under Bolar Exemption only. The following disclaimer applies: These products are offered and sold in small quantities only and solely for uses reasonably related to privileged trials and studies for obtaining marketing authorization required by law (Bolar Exemption). Bachem cannot be made liable for any infringement of intellectual property rights. It is the sole and only responsibility of the purchaser or user of these products to comply with the relevant national rules and regulations.

  • Sequence: Ac-D-2-Nal-D-4-Cpa-D-3-Pal-Ser-N-Me-Tyr-D-Hci-Nle-Arg-Pro-D-Ala-NH₂
  • CAS-No.: 295350-45-7
  • Mol.weight: 1459.11
  • SumFormula: C₇₂H₉₆ClN₁₇O₁₄
  • Bulk item no.: H-7384
  • Synonym(s): D-63153
  • EINECS no.: -
  • MDL no.:

LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 mg H-7384.0001 H-7384.0001BA Each (1mg) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

5 mg H-7384.0005 H-7384.0005BA Each (5mg) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Ozarelix
CAS No. 295350-45-7
Molecular Weight 1459.11
Molecular Formula C₇₂H₉₆ClN₁₇O₁₄
Storage Conditions -20 ± 5 °C
Salt Acetate
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping