Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(ASN7)-AMYLOID B-PROTEIN (1-40), Bachem

Supplier: BACHEM AMERICAS INC
The Tottori (D7N) mutation of β-amyloid peptides accelerates fibrillation without increasing protofibril formation. Ono et al. showed that the English and Tottori mutations alter Abeta assembly at its earliest stages, monomer folding and oligomerization, and produce oligomers that are more toxic to cultured neuronal cells than are wild type oligomers.

  • Sequence: H-Asp-Ala-Glu-Phe-Arg-His-Asn-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH
  • Mol.weight: 4328.88
  • SumFormula: C₁₉₄H₂₉₆N₅₄O₅₇S
  • Bulk item no.: H-7334
  • Synonym(s): Amyloid β-Protein (1-40) Tottori Mutation#Amyloid β-Protein (1-40) D7N



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.5 mg H-7334.0500 H-7334.0500BA Each (1mg) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

1 mg H-7334.1000 H-7334.1000BA Each (1mg) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name (Asn7)-beta-Amyloid (1-40)
Molecular Weight 4328.88
Molecular Formula C₁₉₄H₂₉₆N₅₄O₅₇S
Storage Conditions -20 ± 5 °C
Salt Trifluoroacetate
Peptide (Asn7)-beta-Amyloid (1-40)
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty