Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fmoc-Tyr(Boc-Nmec)-OH, Bachem

Supplier: BACHEM AMERICAS INC
B-4210.0005 B-4210.0001
Fmoc-Tyr derivative for improving the solubility of hydrophobic peptides after cleavage from the resin, as removal of Boc leaves a charged moiety. After purification, the solubilizing Nmec (N-methyl-N-[2-(methylamino)ethyl]carbamoyl) group is split off via an intramolecular cyclization in neutral or slightly basic solution.

  • Sequence: Fmoc-Tyr(Boc-Nmec)-OH
  • CAS-No.: 1033814-21-9
  • Mol.weight: 617.70
  • SumFormula: C₃₄H₃₉N₃O₈
  • Bulk item no.: B-4210
  • Synonym(s): Fmoc-Tyr(Boc-N-methyl-N-[2-(methylamino)ethyl]carbamoyl)-OH
  • EINECS no.: -
  • MDL no.: MFCD22377746

Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 g B-4210.0001 B-4210.0001BA Each (1g) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

5 g B-4210.0005 B-4210.0005BA Each (5g) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Fmoc-Tyr(Boc-Nmec)-Oh
CAS No. 1033814-21-9
Molecular Weight 617.70
Molecular Formula C₃₄H₃₉N₃O₈

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty