Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fmoc-[ring-D₅]Phe-OH, Bachem

Supplier: BACHEM AMERICAS INC
B-4115.1000 B-4115.0500
Fmoc-Phe(D₅)-OH was N-methylated and used in the synthesis of an internal standard for the quantification of the μ-opioid receptor antagonist DAMGO (H-2535) in ovine plasma samples by MALDI-TOF-MS.

  • Sequence: Fmoc-[ring-D₅]Phe-OH
  • CAS-No.: 225918-67-2
  • Mol.weight: 392.47
  • SumFormula: C₂₄H₁₆D₅NO₄
  • Bulk item no.: B-4115
  • Synonym(s):
  • EINECS no.: -
  • MDL no.: MFCD06658797




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
0.5 g B-4115.0500 B-4115.0500BA Each (500mg) Retrieving
1 g B-4115.1000 B-4115.1000BA Each (1g) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Fmoc-(Ring-D5)Phe-Oh
CAS No. 225918-67-2
Molecular Weight 392.47
Molecular Formula C₂₄H₁₆D₅NO₄
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping