(Nle35)-beta-Amyloid25-35 Trifluoroacetate, Bachem
4074464.0001 4074464.0005
H-7314.0001BAEA 116.52 USD
H-7314.0001BA H-7314.0005BA
(Nle35)-beta-Amyloid25-35 Trifluoroacetate, Bachem
Met³⁵ crucially contributes to the oxidative stress associated with the neurotoxic β-amyloid peptides. Aβ 25-35 M35Nle cannot be oxidized, it shows a lower tendency to aggregation and β-turn formation than the WT Aβ.