Etest® Strips, bioMérieux
Supplier: bioMérieux
527458 500958 530358 530058 522558 502658 516058 535058 522658 530258
10500-456PK 548.37 USD
10500-456 10500-444 10500-440 10500-330 10500-408 10500-306 10500-428 10500-390 10500-448 10500-404
Etest® Strips, bioMérieux
Microbiology Test Systems Microorganism Tests
Etest® strips are a stable predefined antibiotic gradient using novel dry chemistry techniques.
  • Precise concentration measurements
  • Cost effective
  • Broadly applicable
Gives a precise Minimum Inhibitory Concentration (MIC) showing single dilution antibiotic gradient with simple interpretation.