Anti-H5N1-H5 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-4813R
10404-664EA 649.69 USD
10404-664
Anti-H5N1-H5 Rabbit Polyclonal Antibody
Antibodies
H5N1-H5

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: H5N1-H5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR