Anti-ABP1 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-4733R
10405-700EA 573.73 USD
10405-700
Anti-ABP1 Rabbit Polyclonal Antibody
Antibodies
ABP1

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR