Anti-plant lectin B4 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-2944R
10352-506EA 573.73 USD
10352-506
Anti-plant lectin B4 Rabbit Polyclonal Antibody
Antibodies
plant lectin B4

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: plant lectin B4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR