Anti-Batroxobin Rabbit Polyclonal Antibody

Supplier: Bioss

BS-2143R
10338-054EA 649.69 USD
10338-054
Anti-Batroxobin Rabbit Polyclonal Antibody
Antibodies
Batroxobin

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Batroxobin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR