Anti-JHDM1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-13601R
10301-744EA 649.69 USD
10301-744
Anti-JHDM1 Rabbit Polyclonal Antibody
Antibodies
KDM2 / JHD1 is a JmjC domain family histone demethylase specific for H3-K36, similar to proteins found in human, mouse, drosophila, X. laevis, C. elegans, and S. pombe.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: JHDM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR