Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SYNGAP Rabbit Polyclonal Antibody

Supplier: Bioss
BS-10392R
The PSD-95/SAP 90 family of proteins, which are known to bind to and cluster various membrane proteins, are involved in the organization of synaptic structure. SynGAP (for synaptic GTPase activating protein), a novel synaptic RasGAP, is a brain-specific protein abundant in the cortex, hippocampus and olfactory bulb. SynGAP interacts with all three PDZ domains within PSD-95/ SAP 90 proteins. It represents one of three classes of mammalian RasGAPs and is specifically localized to excitatory synapses possessing NMDA receptors. SynGAP may be involved in the regulation of BDNF as well as Ras signaling. Its activity is inhibited by phosphorylation by CaMKII, which could result in the activation of the MAP kinase pathway.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SYNGAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg BS-10392R 10243-140 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol SYNGAP
Antigen Name Synaptic Ras GTPase-activating protein 1
Reactivity Mouse, Rat, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms synaptic Ras GTPase activating protein 1 homolog, neuronal RasGAP, MRD5, ras GTPase-activating protein SynGAP, KIAA1938, synaptic Ras GTPase activating protein 1, SYNGAP1, DKFZp761G1421, RASA5, RASA1
Gene ID 8831
Modification Ser836
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty