Anti-VGF/NERP2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-10332R
10365-300EA 649.69 USD
10365-300
Anti-VGF/NERP2 Rabbit Polyclonal Antibody
Antibodies
This gene is specifically expressed in a subpopulation of neuroendocrine cells, and is upregulated by nerve growth factor. The structural organization of this gene is similar to that of the rat gene, and both the translated and the untranslated regions show a high degree of sequence similarity to the rat gene. The encoded secretory protein also shares similarities with the secretogranin/chromogranin family, however, its exact function is not known. Nerve growth factor (NGF) is a peptide that plays a key role in the differentiation and survival of neurons in the peripheral nervous system (PNS) and the central nervous system (CNS). VGF is a peptide synthesized and secreted by neurons and is upregulated by NGF in the PC12 cell line. VGF is widely expressed in both the PNS and CNS, but is especially abundant in the adult hypothalamus. VGF plays an essential role in how the brain regulates energy expenditure and body weight. Its expression is rapidly induced by injury, the circadian clock, and neuronal activity.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: VGF/NERP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR