Anti-STX1A Rabbit Polyclonal Antibody

Supplier: Bioss

BS-10315R
10364-912EA 649.66 USD
10364-912
Anti-STX1A Rabbit Polyclonal Antibody
Antibodies
Syntaxin 1a is potentially involved in docking of synaptic vesicles at presynaptic active zones and may play a critical role in neurotransmitter exocytosis. Haploinsufficiency of STX1A may be the cause of certain cardiovascular and musculo skeletal abnormalities observed in Williams-Beuren syndrome (WBS), a rare developmental disorder.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: STX1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR