Anti-VAV3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-10271R
10364-794EA 649.66 USD
10364-794
Anti-VAV3 Rabbit Polyclonal Antibody
Antibodies
The Vav family of Rho guanine nucleotide exchange factors (GEFs) orchestrate signaling events following lymphocyte antigen receptor activation. Vav3, like Vav (also known as Vav1 or p95Vav), undergoes tyrosine phosphorylation downstream of T cell receptor cross-linkage, and subsequently interacts with 2 adaptor molecules, SLP76 and 3BP2. Following these events, however, the paths of Vav and Vav3 diverge; Vav affects IL-2 promotor activity, while Vav3 impacts gene transcription linked to serum response element (SRE). Furthermore, Vav3 expression follows a cell cycle-dependent pattern, with transient upregulation occuring during mitosis. Encforced Vav3 expression leads to the appearance of multinucleate cells, implicating a role for Vav3 in the control of cytokinesis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: VAV3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR