Anti-RSV Nucleoprotein Rabbit Polyclonal Antibody

Supplier: Bioss

BS-10207R
10363-992EA 649.69 USD
10363-992
Anti-RSV Nucleoprotein Rabbit Polyclonal Antibody
Antibodies
Respiratory syncytial virus (RSV) is a major cause of respiratory illness in young children. RSV infection produces a variety of signs and symptoms involving different areas of the respiratory tract, from the nose to the lungs. RSV is a negative sense, enveloped RNA virus. The virion is variable in shape and size with average diameter of between 120 and 300 nm.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: RSV Nucleoprotein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR