Anti-CHRNA1 Rabbit Polyclonal Antibody

BS-10169R
10363-236EA 271.38 USD
10363-236
Anti-CHRNA1 Rabbit Polyclonal Antibody
Members of the ligand-gated ion channel receptor family are characterized by their fast transmitting response to neurotransmitters. Two important members of this family are the nicotinic acetylcholine and glutamate receptors, both of which are composed of five homologous subunits forming a transmembrane aqueous pore. These transmembrane receptors change conformation in response to their cognate neurotransmitter. Nicotinic acetylcholine receptors (AChRs) are found at the postsynaptic membrane of the neuromuscular junction and bind acetylcholine molecules, allowing ions to move through the pore. Glutamate receptors are found in the postsynaptic membrane of cells in the central nervous system. The activity that is generated at the synapse by the binding of acetylcholine is terminated by acetylcholinesterase, an enzyme that rapidly hydrolyzes acetylcholine. AChR?, also known as ACHRD, CHRNA, CMS2A, FCCMS, SCCMS or CHRNA1, is a 482 amino acid multi-pass membrane protein that exists as two alternatively spliced isoforms, which are expressed in different tissues. Isoform 1 is only expressed in skeletal muscle whereas isoform 2 is constitutively expressed in skeletal muscle, brain, heart, kidney, liver, lung and thymus.

Recommended Dilutions: Western Blot: 1:100-1000; Flow Cytometry: 1:20-100; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CHRNA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR