Anti-CLPS Rabbit Polyclonal Antibody

Supplier: Bioss

BS-10088R
10361-998EA 649.69 USD
10361-998
Anti-CLPS Rabbit Polyclonal Antibody
Antibodies
Colipase (CLPS) is a cofactor of pancreatic lipase and is necessary for efficient dietary lipid hydrolysis. It allows the lipase to anchor itself to the surface of lipid micelles, thereby counteracting the inhibitory effect of intestinal bile salts on the lipase.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR