Anti-AFP4a Rabbit Polyclonal Antibody

Supplier: Bioss

BS-10081R
10361-832EA 649.69 USD
10361-832
Anti-AFP4a Rabbit Polyclonal Antibody
Antibodies
No data available.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: AFP4a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR