|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | NPY |
Antigen Name | Neuropeptide Y |
Reactivity | Mouse, Rat, Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ImmunoChemistry | Yes |
Antigen Synonyms | PYY4 |
Gene ID | 4852 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months. |
Storage Temperature | Store at -20°C for 12 months |
Shipping Temperature | 4°C |