Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CLEC4M Rabbit Polyclonal Antibody

Supplier: Bioss
BS-10064R
Dendritic cells (DC) play a primary role in the immune system as antigen presenting cells. A key molecule, dendritic cell specific ICAM3 grabbing nonintegrin (DC SIGN1), is involved in regulating interactions between DC and resting T cells, including antigen presentation to T cells and enhancement of transinfection of CD4+ T cells by HIV 1. Efforts to identify additional type II membrane proteins resulted in the isolation of a related molecule named CD299, also known as DC SIGNR (DC SIGN Related), DC SIGN2, CD209L, and L SIGN (liver/lymph nodespecific ICAM3 grabbing nonintegrin). CD299 is located on Human chromosome 19p13.3 and shares 73-80% amino acid identity with DC SIGN1. Its structure is similar to DC SIGN1 and therefore binds mannose residues in a calcium dependent fashion, including ICAM3 and HIV 1 gp120. CD299 is polymorphic, since allelic variations of the exon 4 encoded sequence have been isolated. This is further supported by a study demonstrating the ability to isolate a large repertoire of CD299 transcripts largely the result of alternative splicing of the 7 coding exons. CD299 is primarily transcribed in the liver and lymph nodes but not in monocyte derived DC. Expression of CD299 is restricted to endothelial cells derived from liver sinusoids, lymph nodes sinuses, and capillaries, although variable expression in placenta and some monocytic cell lines has also been reported, including both membrane and soluble isoforms of the protein.

Recommended Dilutions: Western Blot: 1:100-1000; Flow Cytometry: 1:20-100; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLEC4M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg BS-10064R 10361-572 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CLEC4M
Antigen Name C-type lectin Domain family 4, member M
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Flow Cytometry Yes
Antigen Synonyms CD209L, DC-SIGNR, LSIGN, CD299, DCSIGNR, DC-SIGN2, HP10347, L-SIGN
Gene ID 10332
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty