Anti-KNDC1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-10056R
10361-352EA 649.69 USD
10361-352
Anti-KNDC1 Rabbit Polyclonal Antibody
Antibodies
KNDC1 is a 1,749 amino acid protein that contains two KIND domains and an N-terminal Ras-GEF domain. Expressed in the cerebral cortex, KNDC1 is a likely guanine nucleotide exchange factor (GEF). Existing as six alternatively spliced isoforms, the gene encoding KNDC1 maps to human chromosome 10q26.3 and mouse chromosome 7 F4. Spanning nearly 135 million base pairs, chromosome 10 makes up approximately 4.5% of total DNA in cells and encodes nearly 1,200 genes. Several protein-coding genes, including those that encode for chemokines, cadherins, excision repair proteins, early growth response factors (Egrs) and fibroblast growth receptors (FGFRs), are located on chromosome 10. Defects in some of the genes that map to chromosome 10 are associated with Charcot-Marie Tooth disease, Jackson-Weiss syndrome, Usher syndrome, nonsyndromatic deafness, Wolman’s syndrome, Cowden syndrome, multiple endocrine neoplasia type 2 and porphyria.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: KNDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR