Anti-PRRSV Rabbit Polyclonal Antibody

Supplier: Bioss
BS-10043R
10361-104EA 573.73 USD
10361-104
Anti-PRRSV Rabbit Polyclonal Antibody
Antibodies
PRRSV

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRRSV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR