Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UEA1 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-10025R
Ulex Europaeus is a European gorse shrub with fragrant golden-yellow flowers. Ulex Europaeus-I Lectin is a 46 kDa glycoprotein known to interact with -L fucosyl residues in oligosaccharides present on the membranes of human blood group O erythrocytes, human endothelial cells and a variety of human and animal epithelial cells.This antibody reacts with Ulex Europaeus Lectin 1 bound to human endothelial cells of normal and neoplastic blood and lymphatic vesicles. It also reacts with human epithelia such as in the colon, bronchus, epidermis, sweat gland ducts and hair follicles. It also reacts with for Ulex Europaeus Lectin 1 bound to vasclular endothelium and squamous epithelium of human tonsil. While erythrocytes may be stained, no other tonsilar elements are reactive with the antibody.This antibody may be useful in the detection of vascular tumors, the This antibody may be useful in the detection of vascular tumors, the investigation of vascular invasion by tumor cells, for determination of Ulex Europaeus Lectin 1 binding to normal, embryonal, dysplastic and neoplastic epithelia and the study of storage diseases such as fucosidosis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: UEA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg BS-10025R 10360-706 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol UEA1
Antigen Name Ulex Europaeus Lectin 1
Reactivity Plant
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms UEA1, UEA I, Ulex europaeus agglutinin; Anti HO lectin 1, UEA 1, UEA-1, Anti HO lectin I., UEAI, UEA-I
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty