Anti-NPY2R Rabbit Polyclonal Antibody

Supplier: Bioss

BS-0937R
10359-376EA 649.69 USD
10359-376
Anti-NPY2R Rabbit Polyclonal Antibody
Antibodies
Receptor for neuropeptide Y and peptide YY. The rank order of affinity of this receptor for pancreatic polypeptides is PYY >NPY >PYY (3-36) >NPY (2-36) >[Ile-31, Gln-34] PP >[Leu-31, Pro-34] NPY >PP, [Pro-34] PYY and NPY free acid.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NPY2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR