Anti-Batroxobin Rabbit Polyclonal Antibody

Supplier: Bioss
BS-0912R
10358-248EA 553.64 USD
10358-248
Anti-Batroxobin Rabbit Polyclonal Antibody
Antibodies
No data available.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Batroxobin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR