Anti-HCMV UL23 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-0896R
10358-640EA 577.94 USD
10358-640
Anti-HCMV UL23 Rabbit Polyclonal Antibody
Antibodies
Human cytomegalovirus (HCMV) is a ubiquitous herpes virus that leads to a life-long persistence. The frequency of infection ranges from 50% to 100% in the general adult population. Human cytomegalovirus causes severe and often fatal disease in immunocompromised individuals including recipients of organ transplants and AIDS patients. It routinely reactivates in healthy virus carriers, but this is usually controlled by the host immune response. Monocytes may be an important reservoir for latent HCMV; however, the primary reservoir may be a more primitive cell from the myeloid lineage. Reactivation may result from cellular differentiation or inflammation.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: HCMV UL23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR