Anti-CSP Rabbit Polyclonal Antibody

Supplier: Bioss

BS-0848R
10357-762EA 649.69 USD
10357-762
Anti-CSP Rabbit Polyclonal Antibody
Antibodies
The circumsporozoite protein is the immunodominant surface antigen on the sporozoite.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CSP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR