Anti-Os05g0477200 protein Rabbit Polyclonal Antibody

Supplier: Bioss

BS-0807R
10357-208EA 649.69 USD
10357-208
Anti-Os05g0477200 protein Rabbit Polyclonal Antibody
Antibodies
Os05g0477200 protein

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Os05g0477200 protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR