Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDK4 Rabbit Polyclonal Antibody

Supplier: Bioss
Pdk4 (pyruvate dehydrogenase kinase, isozyme 4) mediates glucose oxidation through inhibitory phosphorylation of the pyruvate dehydrogenase complex. The pyruvate dehydrogenase complex (PDC) is a mitochondrial matrix enzyme that plays an key role in aerobic energy metabolism by catalyzing the oxidative decarboxylation of pyruvate to form acetyl-CoA for subsequent breakdown in the Krebs cycle. The enzymatic activity of the mammalian PDC is regulated through phosphorylation and desphosphorylation of the E1?subunit of the complex. The enzymes responsible for the phosphorylation and inactivation of the PDC are pyruvate dehydrogenase kinases (PDK1-4). [TISSUE SPECI FICITY] Ubiquitous; highest levels of expression in heart and skeletal muscle. Belongs to the PDK/BCKDK protein kinase family. Contains 1 histidine kinase domain.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PDK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg BS-0682R 10239-868 Each (100µl) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol PDK4
Antigen Name Pyruvate Dehydrogenase Kinase 4
Reactivity Mouse, Rat, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms FLJ40832, Pyruvate dehydrogenase kinase isozyme 4 mitochondrial., Pyruvate dehydrogenase kinase isoform 4, Pyruvate dehydrogenase [lipoamide] kinase isozyme 4 mitochondrial, Pyruvate dehydrogenase kinase 4
Gene ID 5166
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
Storage Temperature Store at -20°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty