Anti-Cdk-associated cullin1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-0614R
10238-542EA 649.69 USD
10238-542
Anti-Cdk-associated cullin1 Rabbit Polyclonal Antibody
Antibodies
Cell cycle associated protein capable of promoting cell proliferation through the activation of CDK2 at the G1/S phase transition.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Cdk-associated cullin1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR