Anti-HAI-1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-0540R
10235-382EA 649.69 USD
10235-382
Anti-HAI-1 Rabbit Polyclonal Antibody
Antibodies
HGFA Inhibitor 1 is an endogenous inhibitor of the kunitz-type serine proteinase HGF-activator (HGFA). HGF was first described as a hepatocyte-specific mitogen and survival factor, and has since been shown to exert a variety of actions on many cell types by binding to its MET receptor. HGF is activated by cleavage of the single-chain form to form a two-chain version by HGFA. HGFA Inhibitor 1 also inhibits trypsin, and MTSP-1 (Matriptase), a serine proteinase that also activates HGF. HGFB Inhibitor 1 is a splice variant of HGFA Inhibitor 1, and contains a 16 amino acid insert after the first kringle domain, relative to the HGFA Inhibitor 1 sequence.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: HAI-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR