Anti-TBX2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-0507R
10235-150EA 649.69 USD
10235-150
Anti-TBX2 Rabbit Polyclonal Antibody
Antibodies
Involved in the transcriptional regulation of genes required for mesoderm differentiation. Probably plays a role in limb pattern formation. Acts as a negative regulator of PML function in cellular senescence. May be required for cardiac atrioventricular canal formation (By similarity).

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TBX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR