Anti-SLC11A2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-0401R
10241-738EA 649.69 USD
10241-738
Anti-SLC11A2 Rabbit Polyclonal Antibody
Antibodies
No data available.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC11A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR