Anti-large S protein Rabbit Polyclonal Antibody

Supplier: Bioss

BS-0343R
10241-228EA 649.69 USD
10241-228
Anti-large S protein Rabbit Polyclonal Antibody
Antibodies
HBV surface proteins [Hepatitis B virus]. Major surface antigen from hepadnavirus; pfam00695.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: large S protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR