Anti-E1 glycoprotein Rabbit Polyclonal Antibody

Supplier: Bioss

BS-0319R
10235-978EA 649.48 USD
10235-978
Anti-E1 glycoprotein Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: E1 glycoprotein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR